Order Entry
Export
ContactUsLinkComponent
Acalabrutinib 98%
Health
Acalabrutinib 98%
Catalog # 472531000.
CAS Number:  
Health
Acalabrutinib 98%
Catalog # 472531000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    white to almost white powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :465.52 g/mol
 :GHS08
Storage Temperature:Refrigerator
CAS Number:1420477-60-6
Formula:C₂₆H₂₃N₇O₂
MDL Number:MFCD29472294