
2,2-Bis(hydroxymethyl)butyric acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10097-02-6
:
Formula:C₆H₁₂O₄
MDL Number:MFCD00190088
:148.16 g/mol
Boiling Pt:360°C
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to almost white crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:97.5 to 102.5 %
- Melting point:110°C to 118°C
- Water:≤0.5 %