
1,4-Dioxan-2-one 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3041-16-5
:
Formula:C₄H₆O₃
MDL Number:MFCD00215855
:102.09 g/mol
Boiling Pt:223°C
Flash Pt:215°C
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to almost white crystalline solid
- Infrared spectrum:Conforms
- GC:≥97.5 %