Y-27632 dihydrochloride 99%
Catalog # 470701000.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:white to beige powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Water:≤8 %
Specification Test Results
About this item
:
:320.26 g/mol
Boiling Pt:258°C (760mmHg)
Storage Temperature:Refrigerator
CAS Number:129830-38-2 | 146986-50-7 (free base)
:146986-50-7
Formula:C₁₄H₂₁N₃O·2HCl
MDL Number:MFCD03490488