
1,4-Diiodotetrafluorobenzene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:392-57-4
:
Formula:C₆F₄I₂
MDL Number:MFCD00001055
:401.87 g/mol
EINECS:206-877-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:white to dark yellow powder or crystals
- Infrared spectrum:Conforms
- GC:≥97.5 %