Order Entry
Export
ContactUsLinkComponent
2-Chloro-p-cresol 97%
Toxic
2-Chloro-p-cresol 97%
Certificates
Toxic
2-Chloro-p-cresol 97%

Specification Test Results

  • Appearance:
    colorless to yellow liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • Refractive index:
    1.5520 to 1.5540 (20°C, 589 nm)

Specification Test Results