



2,4-Dimethylhexane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:589-43-5
:
Formula:C₈H₁₈
MDL Number:MFCD00039929
:114.23 g/mol
Density:0.7
Flash Pt:10°C
Storage Temperature:Ambient
EINECS:209-649-6
UN:1262
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:clear colorless to almost colorless liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Refractive index:1.3940 to 1.3960 (20°C, 589 nm)