Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyrrolo[1,2-a]pyridine
Pyrrolo[1,2-a]pyridine
Pyrrolo[1,2-a]pyridine
:
Thermo Fisher Scientific
Pyrrolo[1,2-a]pyridine
:
Thermo Fisher Scientific
:
274-40-8
:
:
C₈H₇N
:
MFCD00030189
:
117.15 g/mol
:
202°C (760mmHg)
:
GHS07
:
Freezer
Appearance:
white to green-grey crystalline powder
Infrared spectrum:
Conforms
GC:
≥94 %
Melting point:
70°C to 78°C