Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Benzenedimethanethiol

1,4-Benzenedimethanethiol

Irritant
1,4-Benzenedimethanethiol
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  alpha,alpha'-Dimercapto-p-xylene, 1,4-Bis(mercaptomethyl)benzene, 1,4-BDMT
  • CAS Number:105-09-9
  • Formula:C₈H₁₀S₂
  • MDL Number:MFCD00004872
  • Molecular Weight:170.29 g/mol
  • Boiling Pt:156°C (16hPa)
  • Storage Temperature:Ambient
  • EINECS:203-269-4
  • UN:3335
  • ADR:9,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    white powder, crystals or solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • Melting point:
    40°C to 45°C
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 99,10 €
    99,10 €
    1 * 1 g
       99,10 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 349,00 €
    349,00 €
    1 * 5 g
       349,00 €