Order Entry
Export
Orders LinkContactUsLinkComponent
(S)-10-hydroxycamptothecin

(S)-10-hydroxycamptothecin

(S)-10-hydroxycamptothecin
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  1-HCPT, (S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-Hydroxycamptothecin
  • CAS Number:19685-09-7
  • Formula:C₂₀H₁₆N₂O₅
  • MDL Number:MFCD02093100
  • Molecular Weight:364.35 g/mol
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    white to yellow to orange powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Loss on drying:
    ≤5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 109,00 €
    109,00 €
    1 * 1 g
       109,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 354,00 €
    354,00 €
    1 * 5 g
       354,00 €