3-Hydroxyazetidine 95%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 3-Azetidinol, Azetidin-3-ol
- CAS Number:45347-82-8
- Formula:C₃H₇NO
- MDL Number:MFCD03695446
- Molecular Weight:73.09 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: 3-Azetidinol, Azetidin-3-ol
- CAS Number:45347-82-8
- Formula:C₃H₇NO
- MDL Number:MFCD03695446
- Molecular Weight:73.09 g/mol
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:white to grey powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥94 %



