
2-Chloro-4,6-dimethoxypyrimidine 97%
:13223-25-1
:
:C₆H₇ClN₂O₂
:MFCD00274530
:174.58 g/mol
:317°C (760mmHg)
:GHS07
:Ambient
- Appearance:white to light yellow crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Melting point:101°C to 105°C