Sodium alginate, medium viscosity
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9005-38-3
:
MDL Number:MFCD00081310
Merck Index:13,00238
Storage Temperature:Ambient
Specification Test Results
- Appearance:almost white to yellow-brown powder
- Infrared spectrum:Conforms
- Loss on drying:≤15.0 % (100 mg, 105°C, 4 hrs)
- Heavy metals (as Pb):≤20 ppm
- Sulfated ash:30.0 % to 36.0 % (on dried substance)