Acid red 1
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3734-67-6
:
Formula:C₁₈H₁₅N₃O₈S₂
MDL Number:MFCD00003954
:509.41 g/mol
Storage Temperature:Ambient
EINECS:223-098-9
Specification Test Results
- Appearance:red to brown to purple powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Loss on drying:≤10 %
- Specific absorbance A (1%/1cm):≥250 lambda 503 - 509 nm, 0.02 g/l water