Gelonin (from Gelonium multiflorum)
Catalog # ENZOALX350150M001
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 mg
- CAS No.:75037-46-6
- Source:Gelonium multiflorum
- Storage conditions:−80 °C
- UniProtKB:P33186
- Protein/peptide name:Gelonin
- Purity:≥95%
- Molecular weight:~30 kDa
- Concentration:~2 mg/ml
- Formulation:Liquid. In 5 mM phosphate buffer, pH 6,5, containing 5 mM EDTA and 120 mM sodium chloride.
- Shipping temperature:Dry Ice
Specifications
About this item
Type I ribosome inactivating protein (RIP). Depurinates RNA in ribosomes, thus inhibiting protein synthesis in eukaryotic cells, which results in cell death. Widely used to construct immunotoxins composed of cell-targeted antibodies. As a type I RIP it lacks the lectin subunit and is practically non-toxic to intact cells.