Order Entry
Export
ContactUsLinkComponent
HYGOSTAR HACCP Table Dispenser
HYGOSTAR HACCP Table Dispenser
Catalog # MENS88839
Supplier:  MENSCH FRANZ
New Product
HYGOSTAR HACCP Table Dispenser
Catalog # MENS88839
Supplier:  MENSCH FRANZ
New Product

Specifications

  • Colour:
    White/Transparent
  • Description:
    Tissue dispenser, table
  • W×D×H:
    440×340×330 mm
  • Pk:
    1

Specifications

About this item

Plastic table dispenser.

  • Plastic roller stand, transparent/white
  • Suitable for implementing a HACCP system
  • For rolls with a width of up to 30 cm and a roll diameter of up to 25 cm
  • Rubber seals protect against greases, oils, liquids
  • 100% rust-proof, even for humid environments

With PE inner packaging and cardboard outer packaging.