Perfluoropropanesulfonic Acid 50 µg/ml in Methanol:Water
Catalog # EHERA15987234MW-50
Supplier: EHRENSTORFER
New Product
CAS Number:
Specifications
- Pk:1 mL
- CAS No.:423-41-6
- Description:Perfluoropropanesulfonic acid 50 µg/ml in methanol:water
- Hazard class description:Methanol mixture, 3 (6.1), II
- Shipping conditions:20 °C
- UN No.:1230
- GHS signal:Danger
- Formula:C₃HF₇O₃S
- Storage temperature:−18 °C
- Concentration:50 µg/ml
- Molecular weight (Mw):250,09