
Wright's stain for microscopical staining, Sigma-Aldrich®
Catalog # SIAL45252-25G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Density:0.8 g/cm³ (20 °C)
EINECS:273-541-5
:GHS07
Storage Temperature:Ambient
CAS Number:68988-92-1
:51938-72-8
MDL Number:MFCD00082143