Sodium chloride regeneration salt for water softening
Catalog # SGWA01322
Supplier: EVOQUA WATER TECHNOLOGIES
CAS Number:
Specifications
- Pk:2,5 kg
- Seq:4076
Specifications
About this item
:
:58.44 g/mol
Boiling Pt:1416 °C (1013 hPa)
Melting Pt:801 °C
Density:2.17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477