Order Entry
Export
ContactUsLinkComponent
exo-1,3-β-D-Glucanase + β-Glucosidase
exo-1,3-β-D-Glucanase + β-Glucosidase
Catalog # MEGAE-EXBGOS
Supplier:  MEGAZYME
New Product
CAS Number:  
exo-1,3-β-D-Glucanase + β-Glucosidase
Catalog # MEGAE-EXBGOS
Supplier:  MEGAZYME
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • CAS No.:
    exo-1,3-β-glucanase: 9073-49-8
    β-glucosidase: 9001-22-3
  • Source:
    Aspergillus niger
    Trichoderma sp.
  • Enzyme name:
    exo-1,3-β-D-Glucanase + β-Glucosidase
  • Enzyme activity:
    β-Glucosidase, exo-1,3-β-Glucanase
  • Enzyme synonyms:
    glucan 1,3-beta-glucosidase
    3-beta-D-glucan glucohydrolase
    beta-glucosidase
  • Concentration:
    exo-1,3-β-Glucanase (100 U/ml)/β-Glucosidase (20 U/ml)
  • Formulation:
    In 3,2 M ammonium sulphate
  • pH:
    4

Specifications

About this item

High purity exo-1,3-β-D-Glucanase (Trichoderma sp.) + β-Glucosidase (Aspergillus niger) for use in research, biochemical enzyme assays and in vitro diagnostic analysis.

  • Shipping temperature: Ambient
  • Storage temperature: 2 to 8 °C
  • Stability: >1 year under recommended storage conditions
  • CAZy family: GH3, GH55
  • Temperature optima: 40 °C
  • Application: For use in the determination of (1,3)(1,6) β-glucan

A wide range of other carbohydrate active enzymes also available.

Specificity:
exo-1,3-β-glucanase: Successive hydrolysis of β-D-glucose units from the non-reducing ends of (1,3)-β-D-glucans, releasing α-glucose.
β-glucosidase: Hydrolysis of terminal, non-reducing β-D-glucosyl residues with release of β-D-glucose.