Dextran, M.W. ~ 60,000 (from Leuconostoc mesenteroides), BioChemika®, Sigma-Aldrich®
Catalog # SIAL31397-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
EINECS:232-677-5
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935