X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Catalog # ROCH10651745001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:408.63 g/mol
Melting Pt:237…239 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
EINECS:230-640-8
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666