
Chromotropic acid disodium salt dihydrate
Catalog # MOLE25454876-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:400.29 g/mol
Merck Index:13,02261
Melting Pt:300 °C
EINECS:204-972-9
:GHS07
Storage Temperature:Ambient
CAS Number:5808-22-0
Formula:(HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MDL Number:MFCD00150612