Order Entry
Export
ContactUsLinkComponent
 
Irritant
Nitro blue tetrazolium chloride (NBT)
Catalog # ROCH11585029001
Supplier:  ROCHE
CAS Number:  
Irritant
Nitro blue tetrazolium chloride (NBT)
Catalog # ROCH11585029001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :817.65 g/mol
Melting Pt:200 °C
 :M-Clarity™ quality level = MQ100
EINECS:206-067-4
 :GHS07
Storage Temperature:Refrigerator
CAS Number:298-83-9
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159