Order Entry
Export
ContactUsLinkComponent
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS), Sigma-Aldrich®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS), Sigma-Aldrich®
Catalog # ROCH10810118001
Supplier:  ROCHE
CAS Number:  
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS), Sigma-Aldrich®
Catalog # ROCH10810118001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :614.89 g/mol
Merck Index:13,02052
Melting Pt:116 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:75621-03-3
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083