Order Entry
Export
ContactUsLinkComponent
N,N-Diethyl-p-phenylenediammonium sulphate
Irritant
N,N-Diethyl-p-phenylenediammonium sulphate
Catalog # MOLEM62511832
Supplier:  Molekula
CAS Number:  
Irritant
N,N-Diethyl-p-phenylenediammonium sulphate
Catalog # MOLEM62511832
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :262.33 g/mol
Boiling Pt:274…275 °C (1013 hPa)
Melting Pt:184…186 °C
EINECS:228-500-6
 :GHS07
Storage Temperature:Ambient
CAS Number:6283-63-2
Formula:C₁₀H₁₈N₂O₄S
MDL Number:MFCD00012993