
N,N-Dimethyl-1,3-phenylenediammonium dichloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3575-32-4
:
Formula:C₈H₁₄Cl₂N₂
MDL Number:MFCD00012971
:209.12 g/mol
Melting Pt:217 °C (dec.)
Storage Temperature:Ambient
EINECS:222-693-0
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White to beige to grey to purple Powder or crystals
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %