1,3-Di-tert-butylbenzene, Sigma-Aldrich®
Catalog # SIAL272051-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.33
:M-Clarity™ quality level = MQ100
CAS Number:1014-60-4
Formula:C₁₄H₂₂
MDL Number:MFCD00008830