1,3-Di-tert-butylbenzene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1014-60-4
:
Formula:C₁₄H₂₂
MDL Number:MFCD00008830
:190.33
Boiling Pt:106…107 °C (18 mmHg)
Melting Pt:10…11 °C
Density:0.859
Flash Pt:83 °C (183 °F)
Storage Temperature:Ambient