Orders LinkContactUsLinkComponent
VWR® Miscellaneous Dissolution Accessories
VWR® Miscellaneous Dissolution Accessories
Ever-clear dissolution water bath preservative, single dose, 12 pack
Catalog # VWRU75856-092
Supplier:   VWR Collection
VWR® Miscellaneous Dissolution Accessories
Catalog # VWRU75856-092
Supplier:  VWR Collection
VWR® Miscellaneous Dissolution Accessories
VWR® Miscellaneous Dissolution Accessories
Ever-clear dissolution water bath preservative, single dose, 12 pack
Price
438,00 €
Log in for availability
1 * 12 items
Product Details & Documents
Everclear water bath preservative is a crystalline formula offered in premeasured vials for use with dissolution water baths. Use with DEIONIZED water.
The following products are compatible with Agilent/VanKel equipment: APP7 accessories, rotating bottle accessories, Auto-sampler accessories and Paper Rolls.
PTFE suspension cups can be used with most dissolution baths.
A single dose will protect a bath for up to 8 weeks.
Specifications
  • Description:
    Ever-clear dissolution water bath preservative, single dose, 12 pack