Order Entry
Export
ContactUsLinkComponent
Softaskin® pure Wash Lotion for Very Sensitive Skin
Softaskin® pure Wash Lotion for Very Sensitive Skin
Catalog # BRAU180206
Supplier:  B. Braun
New Product
Softaskin® pure Wash Lotion for Very Sensitive Skin
Catalog # BRAU180206
Supplier:  B. Braun
New Product

Specifications

  • Packed:
    500 ml dispenser bottle
  • Description:
    Wash lotion
  • Pk:
    20

Specifications

About this item

Low-allergen wash lotion with skin-neutral pH value.

  • Suitable as a shower gel or for baths, especially for sensitive and exposed skin
  • Free from microplastics and parabens
  • Contains high-quality surfactants and is biodegradable
  • Perfume and colorant free
  • Dermatologically tested

Application: For hand washing in all medical and other facilities especially in neonatology,
oncology, dermatology, intensive care units, and kitchen areas, for patient washing prior to surgery and incontinence patients.