Order Entry
Export
ContactUsLinkComponent
OPTI-LYNX™ Micro Trap Cartridges
OPTI-LYNX™ Micro Trap Cartridges
Catalog # OPTT11-04755-TD
OPTI-LYNX™ Micro Trap Cartridges
Catalog # OPTT11-04755-TD

Specifications

  • Column Format:
    Capillary
  • Flow rate:
    40 - 160 µ l/min
  • Ø int.:
    1 mm
  • Length:
    5 mm
  • Seq:
    01
  • Phase:
    C18AQ
  • Bed dimensions:
    4 µl
  • Pk:
    5

Specifications

About this item

Combining a versatile selection of packed beds and the use of a convenient quick-connect holder, OPTI-LYNX™ provides numerous options for chromatographers for on-line or off-line sample clean-up and pre-concentration.

  • Quick-connect system in-line (BC) or direct-connect versions
  • 414 bar with hand-tight quarter turn, no tools needed
  • Low back pressure design
  • Bi-directional
  • Biocompatible cartridges
  • Available in packages of five cartridges

Two styles of holders are available: The Opti-Lynx Micro BC offers full biocompatibility as the in-line version while the direct-connect configuration threads conveniently into all 10-32 ports.

OPTI-LYNX™ traps are the ideal tool for optimising your technique, whether you want to separate a peptide digest from its matrix for further analyses, or prepare a dilute small molecule sample for LC injection without loss of sample. These columns may be loaded and regenerated repeatedly for maximum value.