Flask, Iodine
Catalog # WEPA019432
Supplier: WEPA PUALUS AND THEWALT
New Product
Specifications
- Capacity:250 ml
- Pk:1
Specifications
About this item
Iodine flask with NS, wide neck made of borosilicate glass, with NS and lid stopper.
- Standard ground joint: 45/40