Orders LinkContactUsLinkComponent
GloCell™ Fixable Viability Dye UV 450
GloCell™ Fixable Viability Dye UV 450
GloCell™ Fixable Viability Dye UV 450
Certificates
GloCell™ Fixable Viability Dye UV 450
GloCell™ Fixable Viability Dye UV 450
2 Options Available Below
Product Details & Documents
Amine-labelling fluorescent dye for live/dead staining of mammalian cells - 100 tests.
GloCell™ Fixable viability dye UV 450 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be utilised prior to fixation, permeabilisation, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay.
GloCell™ Fixable viability dye UV 450 is excited by the UV laser at 355 nm and it has a fluorescence emission maximum of 450 nm that can be detected using the 450/50 band pass filter used to detect DAPI, Hoechst 33258, etc.
CAS No: 67-68-5
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
Product Information
  • Pk Info
    Pk
    Availability
    Price
  • STMC75008.1
    GloCell™ Fixable viability dye UV 450
    GloCell™ Fixable viability dye UV 450
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 184,00 €
    184,00 €
    1 * 100 Tests
       184,00 €
  • STMC75008
    GloCell™ Fixable viability dye UV 450
    GloCell™ Fixable viability dye UV 450
    500 tests
    500 Tests
    Log in for availability
    Log in for availability
    1 * 500 Tests
    1 * 500 Tests - 710,00 €
    710,00 €
    1 * 500 Tests
       710,00 €