Order Entry
Export
ContactUsLinkComponent
Tetramethylrhodamine ethyl ester perchlorate (TMRE) fluorescent dye
Tetramethylrhodamine ethyl ester perchlorate (TMRE) fluorescent dye
Catalog # STMC100-0992
CAS Number:  
undefined
Tetramethylrhodamine ethyl ester perchlorate (TMRE) fluorescent dye
Catalog # STMC100-0992
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :514.95 g/mol
CAS Number:115532-52-0
Formula:C₂₆H₂₇ClN₂O₇
MDL Number:MFCD00467973