Order Entry
Export
Orders LinkContactUsLinkComponent
Carmine alum solution for dye

Carmine alum solution for dye

100 mL
Catalog # STMC07070
Carmine alum solution for dye
Catalog # STMC07070
CAS Number:  
Carmine alum solution for dye
Carmine alum solution for dye

100 mL

CAS Number:  
Price
857,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Dye for staining mouse mammary gland whole mounts.

Carmine Alum is used for the whole mount staining of mouse mammary gland.

Contains
Carmine and aluminum potassium sulfate in water
ProClin™ is added as a preservative

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 mL

Specifications