GemCell™ Plus Xeno-Free Human Serum, Aseptic Assurance System
GEMB100-912-750
New Product
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:GemCell™ Plus xeno-free, human serum AB - AAS
- Environmentally Preferable:
- Shipping conditions:Ships on dry ice
- Origin:US
- Storage temperature:Store frozen at −20...−10 °C
- Pk Info:200×100 ml
- Pack type:BAGA
- Pk:100 mL
Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free human serum is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material goes through additional rigorous testing and contains therapeutic grade human thrombin. All donor units are tested for viral markers and found to be non-reactive.
- This product was developed without the use of animal material
- Custom fill sizes in bottle and bag format offered
- 0,1 µm sterile-filtered
- Supports clinical and commerical applications
- Aseptic Assurance System