Black Ink Cartridge for BradyJet™ J7300 Colour Label Printer, K True
BRDY176505
: Brady
- For Use With:BradyJet™ J7300 colour label printer
- Colour:Black
- Description:K True black ink cartridge, 40,64×71,12×99,06 mm
- Pk:1
Get better results with BradyJet™ J7300 inks. Designed specifically for the J7300, BradyJet™ ink cartridges produce vibrant colours, last long, and deliver value in every drop.
- K True Black ink cartridge delivers a deep black, darker tones, reliable barcode readability, and sharp text
- Works in tandem with separate CMY ink cartridge to mix more accurate, durable colours
- For use with the BradyJet™ J7300 colour label printer
Ink works with enhanced J7300 label material topcoat to dry instantly and won’t smudge, smear, or spot. Self-contained cartridge drops into the printer with a click. Designed to hold up to 50% more ink for longer use, fewer changeovers, and less cartridge waste.