Order Entry
Export
ContactUsLinkComponent
Ink Cartridge for BradyJet™ J7300 Colour Label Printer, CMY
Ink Cartridge for BradyJet™ J7300 Colour Label Printer, CMY
Catalog # BRDY176504
Supplier:  Brady
undefined
Ink Cartridge for BradyJet™ J7300 Colour Label Printer, CMY
Catalog # BRDY176504
Supplier:  Brady

Specifications

  • For Use With:
    BradyJet™ J7300 colour label printer
  • Colour:
    Cyan/magenta/yellow
  • Description:
    CMY ink cartridge, 40,64×71,12×99,06 mm
  • Pk:
    1

Specifications

About this item

Get better results with BradyJet™ J7300 inks. Designed specifically for the J7300, BradyJet™ ink cartridges produce vibrant colours, last long, and deliver value in every drop.

  • CMY ink cartridge delivers vibrant, full-colour labels that demand attention
  • Works in tandem with separate K True Black ink cartridge to mix more accurate, durable colours
  • For use with the BradyJet™ J7300 colour label printer

Ink works with enhanced J7300 label material topcoat to dry instantly and won’t smudge, smear, or spot. Self-contained cartridge drops into the printer with a click. Designed to hold up to 50% more ink for longer use, fewer changeovers, and less cartridge waste.