ContactUsLinkComponent
Overboots, Antistatic
Overboots, Antistatic
Catalog # SSSA000607-W-43-48
Supplier:  3S ARBEITSSCHUTZ
New Product
Overboots, Antistatic
Catalog # SSSA000607-W-43-48
Supplier:  3S ARBEITSSCHUTZ
New Product
Specifications
  • Shoe size:
    43 - 48
  • Colour:
    White
  • Pk:
    600
Specifications
About this item
Tyvek is a lightweight material with high tear and abrasion resistance. It consists of fine, spun continuous polyethylene fibres that are compressed into a fleece-like material.
  • Non-slip sole
  • Height: ~570 mm
The air and water vapor permeability promises good wearing comfort and the material offers high protection against airborne particles and numerous water-soluble inorganic chemicals.