ContactUsLinkComponent
Buccutite™ Rapid Oligo Antibody Conjugation Kit
Buccutite™ Rapid Oligo Antibody Conjugation Kit
Catalog # AATB5450
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Buccutite™ Rapid Oligo Antibody Conjugation Kit
Catalog # AATB5450
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    2
  • Includes:
    2 labellings
Specifications
About this item
Optimised for labelling 100 µg protein. Buccutite™ rapid oligo antibody conjugation kit is optimised for conjugating 100 µg protein with an amino-modified oligo.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped at RT.
Caution: Research use only.