Alconox® Detergent, Powder
SIALZ742914-1.8KG
: Merck
New Product
- Description:Alconox® detergent, powder
- Pack type:Cartons
- Pk:1,8 kg
ALCONOX® is a mixture of a homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, and carbonates. Powdered detergent for general lab cleaning.
- Concentrated and long-lasting
- Biodegradable and readily disposable
- Penetrating wetting power to save
- Alternative for corrosive acids and hazardous solvents
- Easy to rinse, doesn't form interfering residues, and provides reliable results