

Acetone reagent ≥99.5% ACS
SIAL320110-1L
: Merck
New Product
:
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Appearance (Colour):Colourless
- Appearance (Form):Liquid
- Infrared Spectrum:Conforms to Structure
- Colour Test:≤10 APHA
- Purity (GC)
Methanol Content:≤0.05% - GC Identified Peak
Isopropyl Alcohol:≤0.05% - Corrected for Water Content
Purity by GC:≥99.50% - Residue on Evaporation:≤0.001%
- Water (by Karl Fischer):≤0.5%
- Titrable Acid (meq/g) < 0.0003:≤0.0003
- Titrable Base (meq/g):≤0.0006
- Aldehyde (as Formaldehyde):≤0.002%
- Substances reducing MnO4:Pass
- Solubility:Pass
- Meets ACS Requirements:Pass
- Recommended Retest Period:Current ACS Specification
2 Years
:
:58.08 g/mol
:56 °C/760 mmHg (lit.)
:–94 °C (lit.)
:0.791 g/ml (25 °C) (lit.)
:−17.0 °C
:1090
:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:67-64-1
:CH₃COCH₃
:MFCD00008765