0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Trifluoromethyl phenyl diazirine amine
Trifluoromethyl phenyl diazirine amine
Trifluoromethyl phenyl diazirine amine
Trifluoromethyl phenyl diazirine amine
Catalog # AATB39009
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
Trifluoromethyl phenyl diazirine amine
Catalog # AATB39009
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
Product Details