Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8WS amine [TQ8WS amine]
8WS amine [TQ8WS amine], Tide Quencher™
8WS amine [TQ8WS amine], Tide Quencher™
8WS amine [TQ8WS amine], Tide Quencher™
Catalog # AATB2131
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
8WS amine [TQ8WS amine], Tide Quencher™
Catalog # AATB2131
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1034.01 g/mol
Product Details