0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
7.2WS maleimide [TQ7.2WS maleimide]
7.2WS maleimide [TQ7.2WS maleimide], Tide Quencher™
7.2WS maleimide [TQ7.2WS maleimide], Tide Quencher™
7.2WS maleimide [TQ7.2WS maleimide], Tide Quencher™
Catalog # AATB2126
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
7.2WS maleimide [TQ7.2WS maleimide], Tide Quencher™
Catalog # AATB2126
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1100.16 g/mol
Product Details