0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
7.1WS azide [TQ7.1WS azide]
7.1WS azide [TQ7.1WS azide], Tide Quencher™
7.1WS azide [TQ7.1WS azide], Tide Quencher™
Catalog # AATB2120
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
7.1WS azide [TQ7.1WS azide], Tide Quencher™
Catalog # AATB2120
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1046.12 g/mol
Product Details