0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
iFluor® 605 succinimidyl ester
iFluor® 605 succinimidyl ester
iFluor® 605 succinimidyl ester
iFluor® 605 succinimidyl ester
Catalog # AATB1420
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
iFluor® 605 succinimidyl ester
Catalog # AATB1420
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1004.22 g/mol
Product Details