Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fluorescein tyramide

1 mg
Catalog # AATB11062
Supplier:   AAT Bioquest
Fluorescein tyramide
Catalog # AATB11062
Supplier:  AAT Bioquest
CAS Number:  210236-90-1
Fluorescein tyramide

1 mg

CAS Number:  210236-90-1
Price
288,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Fluorescein Tyramide is a green fluorescent reagent widely used for tyramide signal amplification (TSA) in IHC, ICC, FISH and multicolor FISH.

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • CAS Number:210236-90-1
  • Molecular Weight:495.49 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications