0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde [5-FAM aldehyde]
Catalog # AATB9001
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
Fluorescein aldehyde [5-FAM aldehyde]
Catalog # AATB9001
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
550.52 g/mol
Product Details